ADVH49B82F9E AChemBlock

(1R)-1-(oxetan-3-yl)ethanol

  • Molecular Weight : 102.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-1-oxetan-3-yl-ethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86193
Availability details for
Quantity
  • Availability
  • Add to Cart