ADVH49B82FA5 AChemBlock

(1,4,4-trifluorocyclohexyl)methanamine

  • Molecular Weight : 167.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-4-trifluorocyclohexyl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86200
Availability details for
Quantity
  • Availability
  • Add to Cart