ADVH49B82FA7 AChemBlock

3-amino-1-(4-methoxybenzyl)pyrrolidin-2-one

  • Cas Number : 1248990-38-6
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-1-4-methoxybenzyl-pyrrolidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70253
Availability details for
Quantity
  • Availability
  • Add to Cart