ADVH49B82FAA AChemBlock

7-chloro-3-nitroquinolin-4-ol

  • Cas Number : 5350-50-5
  • Molecular Weight : 224.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-3-nitroquinolin-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70254
Availability details for
Quantity
  • Availability
  • Add to Cart