ADVH49B82FAD AChemBlock

tert-butyl ((1R,3S)-3-iodocyclopentyl)carbamate

  • Cas Number : 1932119-33-9
  • Molecular Weight : 311.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1r-3s-3-iodocyclopentyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70256
Availability details for
Quantity
  • Availability
  • Add to Cart