ADVH49B82FAF AChemBlock

N-(5-bromo-2-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide

  • Cas Number : 2055538-86-6
  • Molecular Weight : 398.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-5-bromo-2-fluorophenyl-3-trifluoromethyl-benzenesulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70258
Availability details for
Quantity
  • Availability
  • Add to Cart