ADVH49B82FB2 AChemBlock

phenyl bis((2-chloroethyl)-l2-azaneyl)phosphinate

  • Molecular Weight : 295.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/phenyl-bis-2-chloroethyl-l2-azaneyl-phosphinate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70261
Availability details for
Quantity
  • Availability
  • Add to Cart