ADVH49B82FB5 AChemBlock

2-amino-N-(benzyloxy)benzamide

  • Cas Number : 4797-75-5
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-n-benzyloxy-benzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70264
Availability details for
Quantity
  • Availability
  • Add to Cart