ADVH49B82FB9 AChemBlock

3-bromo-4-nitroquinoline

  • Cas Number : 872826-82-9
  • Molecular Weight : 253.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-nitroquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70268
Availability details for
Quantity
  • Availability
  • Add to Cart