ADVH49B82FBF AChemBlock

4-benzyl-2-chloropyrimidine

  • Cas Number : 168150-96-7
  • Molecular Weight : 204.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-benzyl-2-chloropyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70275
Availability details for
Quantity
  • Availability
  • Add to Cart