ADVH49B82FC6 AChemBlock

2-(4-(ethoxycarbonyl)phenyl)acetic acid

  • Cas Number : 67097-50-1
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-ethoxycarbonyl-phenyl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70282
Availability details for
Quantity
  • Availability
  • Add to Cart