ADVH49B82FC9 AChemBlock

methyl 4-(((2-chloroquinazolin-4-yl)amino)methyl)benzoate

  • Molecular Weight : 327.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-2-chloroquinazolin-4-yl-amino-methyl-benzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70287
Availability details for
Quantity
  • Availability
  • Add to Cart