ADVH49B82FCC AChemBlock

2-(4'-methoxy-1,2,3,6-tetrahydro-[1,1'-biphenyl]-4-yl)-4,4,5-trimethyl-1,3,2-dioxaborolane

  • Molecular Weight : 300.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-methoxy-1-2-3-6-tetrahydro-1-1-biphenyl-4-yl-4-4-5-trimethyl-1-3-2-dioxaborolane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70290
Availability details for
Quantity
  • Availability
  • Add to Cart