ADVH49B82FDE AChemBlock

4-bromoisothiazole-3-carbaldehyde

  • Molecular Weight : 192.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromoisothiazole-3-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71975
Availability details for
Quantity
  • Availability
  • Add to Cart