ADVH49B82FF6 AChemBlock

(5-fluoroisoquinolin-8-yl)methanol

  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-fluoroisoquinolin-8-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72063
Availability details for
Quantity
  • Availability
  • Add to Cart