ADVH49B82FF7 AChemBlock

2-(4-(trifluoromethyl)phenyl)cyclopropanecarboxylic acid

  • Molecular Weight : 230.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-trifluoromethyl-phenyl-cyclopropanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72447
Availability details for
Quantity
  • Availability
  • Add to Cart