ADVH49B83001 AChemBlock

methyl 5-bromo-2,3-difluorobenzoate

  • Molecular Weight : 251.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-2-3-difluorobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72128
Availability details for
Quantity
  • Availability
  • Add to Cart