ADVH49B83017 AChemBlock

3-Bromo-6-fluoro-2-cyanophenol

  • Molecular Weight : 216.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-6-fluoro-2-cyanophenol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72188
Availability details for
Quantity
  • Availability
  • Add to Cart