ADVH49B83019 AChemBlock

4-Amino-3-bromo-6-fluoroanisole

  • Molecular Weight : 220.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-amino-3-bromo-6-fluoroanisole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72191
Availability details for
Quantity
  • Availability
  • Add to Cart