ADVH49B8301E AChemBlock

1,2-Diamino-4-(difluoromethoxy)benzene

  • Molecular Weight : 174.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-diamino-4-difluoromethoxy-benzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72197
Availability details for
Quantity
  • Availability
  • Add to Cart