ADVH49B83020 AChemBlock

3,4-diaminophenylsulfonyl methane

  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-diaminophenylsulfonyl-methane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72199
Availability details for
Quantity
  • Availability
  • Add to Cart