ADVH49B83024 AChemBlock

1,2-Diamino-3-iodo-5-fluorobenzene

  • Molecular Weight : 252.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-diamino-3-iodo-5-fluorobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72203
Availability details for
Quantity
  • Availability
  • Add to Cart