ADVH49B8302C AChemBlock

(3-methylisoquinolin-7-yl)methanol

  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methylisoquinolin-7-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72211
Availability details for
Quantity
  • Availability
  • Add to Cart