ADVH49B83037 AChemBlock

(8-fluoroisoquinolin-5-yl)methanamine

  • Molecular Weight : 176.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-fluoroisoquinolin-5-yl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72229
Availability details for
Quantity
  • Availability
  • Add to Cart