ADVH49B83052 AChemBlock

6-Bromo-5-fluoro-1H-benzimidazole

  • Molecular Weight : 215.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-5-fluoro-1h-benzimidazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72284
Availability details for
Quantity
  • Availability
  • Add to Cart