ADVH49B83053 AChemBlock

methyl 4-(((methylsulfonyl)oxy)methyl)bicyclo[2.2.2]octane-1-carboxylate

  • Molecular Weight : 276.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-methylsulfonyl-oxy-methyl-bicyclo-2-2-2-octane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72285
Availability details for
Quantity
  • Availability
  • Add to Cart