ADVH49B83054 AChemBlock

5-bromo-6-iodo-1H-indazole

  • Molecular Weight : 322.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-iodo-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72286
Availability details for
Quantity
  • Availability
  • Add to Cart