ADVH49B8305C AChemBlock

1-(4-hydroxyphenyl)piperazin-2-one

  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-hydroxyphenyl-piperazin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72316
Availability details for
Quantity
  • Availability
  • Add to Cart