ADVH49B83061 AChemBlock

Seviteronel enantiomer

  • Molecular Weight : 399.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/seviteronel-enantiomer.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84832
Availability details for
Quantity
  • Availability
  • Add to Cart