ADVH49B8306C AChemBlock

tert-butyl 2-(6-bromo-1-(4-methoxybenzyl)-5-methyl-2,4-dioxo-1,4-dihydrothieno[2,3-d]pyrimidin-3(2H)-yl)-2-methylpropanoate

  • Molecular Weight : 523.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-6-bromo-1-4-methoxybenzyl-5-methyl-2-4-dioxo-1-4-dihydrothieno-2-3-d-pyrimidin-3-2h-yl-2-methylpropanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86460
Availability details for
Quantity
  • Availability
  • Add to Cart