ADVH49B83070 AChemBlock

GSK963 racemate

  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/gsk963-racemate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86781
Availability details for
Quantity
  • Availability
  • Add to Cart