ADVH49B83072 AChemBlock

methyl (S)-3-(8-bromo-1-methyl-6-(pyridin-2-yl)-4H-benzo[f]imidazo[1,2-a][1,4]diazepin-4-yl)propanoate benzenesulfonate

  • Cas Number : 1001415-66-2
  • Molecular Weight : 597.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-s-3-8-bromo-1-methyl-6-pyridin-2-yl-4h-benzo-f-imidazo-1-2-a-1-4-diazepin-4-yl-propanoate-benzenesulfonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84261
Availability details for
Quantity
  • Availability
  • Add to Cart