ADVH49B83075 AChemBlock

MKC-3946

  • Cas Number : 1093119-54-0
  • Molecular Weight : 380.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/mkc-3946.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72386
Availability details for
Quantity
  • Availability
  • Add to Cart