ADVH49B83076 AChemBlock

tributyl(3-methoxyprop-1-yn-1-yl)stannane

  • Cas Number : 113794-24-4
  • Molecular Weight : 359.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tributyl-3-methoxyprop-1-yn-1-yl-stannane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72348
Availability details for
Quantity
  • Availability
  • Add to Cart