ADVH49B83077 AChemBlock

(R)-6-((4-chlorophenyl)(hydroxy)(1-methyl-1H-imidazol-5-yl)methyl)-4-(3-ethynylphenyl)-1-methylquinolin-2(1H)-one

  • Cas Number : 1190094-64-4
  • Molecular Weight : 479.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/r-6-4-chlorophenyl-hydroxy-1-methyl-1h-imidazol-5-yl-methyl-4-3-ethynylphenyl-1-methylquinolin-2-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72385
Availability details for
Quantity
  • Availability
  • Add to Cart