ADVH49B8307A AChemBlock

AG99

  • Cas Number : 122520-85-8
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/ag99.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86724
Availability details for
Quantity
  • Availability
  • Add to Cart