ADVH49B8307B AChemBlock

ERK5-IN-1

  • Cas Number : 1234479-76-5
  • Molecular Weight : 459.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/erk5-in-1-310002.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86692
Availability details for
Quantity
  • Availability
  • Add to Cart