ADVH49B83084 AChemBlock

5-(tributylstannyl)isothiazole

  • Cas Number : 1416853-22-9
  • Molecular Weight : 374.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-tributylstannyl-isothiazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72340
Availability details for
Quantity
  • Availability
  • Add to Cart