ADVH49B83087 AChemBlock

Ivosidenib

  • Cas Number : 1448347-49-6
  • Molecular Weight : 582.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/ivosidenib.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86637
Availability details for
Quantity
  • Availability
  • Add to Cart