ADVH49B8308D AChemBlock

tert-Butyl(1-aminopropan-2-yl)(tributylstannyl)methyl)carbamate(SnAP-2-Me-pip)

  • Cas Number : 1557288-01-3
  • Molecular Weight : 477.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-aminopropan-2-yl-tributylstannyl-methyl-carbamate-snap-2-me-pip.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72338
Availability details for
Quantity
  • Availability
  • Add to Cart