ADVH49B83091 AChemBlock

RAF-709

  • Cas Number : 1628838-42-5
  • Molecular Weight : 542.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/raf-709.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72373
Availability details for
Quantity
  • Availability
  • Add to Cart