ADVH49B830A9 AChemBlock

(1r,4r)-4-(2-methoxyethoxy)cyclohexan-1-amine hydrochloride

  • Molecular Weight : 209.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-4-2-methoxyethoxy-cyclohexan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84371
Availability details for
Quantity
  • Availability
  • Add to Cart