ADVH49B830AD AChemBlock

O5-tert-butyl O3-ethyl spiro[4,6-dihydro-2H-pyrazolo[4,3-c]pyridine-7,1'-cyclopropane]-3,5-dicarboxylate

  • Molecular Weight : 321.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/o5-tert-butyl-o3-ethyl-spiro-4-6-dihydro-2h-pyrazolo-4-3-c-pyridine-7-1-apos-cyclopropane-3-5-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84847
Availability details for
Quantity
  • Availability
  • Add to Cart