ADVH49B830AF AChemBlock

methyl (Ra)-2-bromospiro[3.3]heptane-6-carboxylate

  • Molecular Weight : 233.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-ra-2-bromospiro-3-3-heptane-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84969
Availability details for
Quantity
  • Availability
  • Add to Cart