ADVH49B830B4 AChemBlock

D-Methionine sulfoxide hydrochloride

  • Molecular Weight : 201.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/d-methionine-sulfoxide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K16158
Availability details for
Quantity
  • Availability
  • Add to Cart