ADVH49B830B7 AChemBlock

1-(1-methyl-1h-benzoimidazol-2-yl)-piperidine-4-carboxylic acid hydrochloride

  • Molecular Weight : 295.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/1-1-methyl-1h-benzoimidazol-2-yl-piperidine-4-carboxylic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K16180
Availability details for
Quantity
  • Availability
  • Add to Cart