ADVH49B830BC AChemBlock

6-bromo-4-chloro-2H-indol-2-one

  • Molecular Weight : 244.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-4-chloro-2h-indol-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87304
Availability details for
Quantity
  • Availability
  • Add to Cart