ADVH49B830BD AChemBlock

(2-(tert-butoxycarbonyl)naphthalen-1-yl)glycine

  • Cas Number : 146621-93-4
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-tert-butoxycarbonyl-naphthalen-1-yl-glycine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87641
Availability details for
Quantity
  • Availability
  • Add to Cart