ADVH49B830BE AChemBlock

1-(4-chloro-2-fluorophenyl)ethan-1-amine acetate hydrochloride

  • Cas Number : 856562-91-9
  • Molecular Weight : 270.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-chloro-2-fluorophenyl-ethan-1-amine-acetate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88032
Availability details for
Quantity
  • Availability
  • Add to Cart