ADVH49B830BF AChemBlock

chloridazone-desphenyl hydrochloride

  • Cas Number : 6339-19-1
  • Molecular Weight : 145.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/chloridazone-desphenyl-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72323
Availability details for
Quantity
  • Availability
  • Add to Cart